Thermo Scientific™ Hypersil™ Classical Silica HPLC Columns
High-efficiency columns with proven results and reproducibility
  • Suitable for Silica phase
Thermo Scientific™ Hypersil™ columns have been trusted by researchers for over 30 years. Each batch of silica produced for these columns is tested to make sure it results in the same separations as previous batches, guaranteeing you have the same results time after time, even after changing your column. The wide range of phases ensures a column is available to match your application.
Hypersil Silica Columns are produced using an unbonded media that has a narrow particle size distribution, providing high performance and efficiency, as well as excellent batch-to-batch reproducibility. These columns are useful for the separation of non-polar and moderately polar compounds in normal phase separations. Stable in pH range of 2 to 8.
297,000.00 - ₹358,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific 30005-054630 HPLC Column, Silica, 5 µm, 50 mm L x 4.6 mm ID 0188704 Mfr # 30005-054630
COMPARE
4.6 50 5
₹297,000.00INR / Each
Thermo Scientific Hypersil 30005-154030 HPLC Column, Silica, 5 µm, 150 mm L x 4.0 mm ID 0188711 Mfr # 30005-154030
COMPARE
4 150 5
₹342,000.00INR / Each
Thermo Scientific Hypersil 30005-254630 HPLC Column, Silica, 5 µm, 250 mm L x 4.6 mm ID 0188716 Mfr # 30005-254630
COMPARE
4.6 250 5
₹358,000.00INR / Each