Thermo Scientific™ Hypersil APS-2 (Amino) Classical HPLC Columns
High-efficiency columns with proven results and reproducibility
Thermo Scientific Hypersil columns have been trusted by researchers for over 30 years. Each batch of silica produced for these columns is tested to make sure it results in the same separations as previous batches, guaranteeing you have the same results time after time, even after changing your column. The wide range of phases ensures a column is available to match your application.
Hypersil APS-2 (Amino) Columns provide a versatile amino propyl phase and are extra sensitive for the analysis of sugars. These columns may be used for reversed phase, normal phase, and weak anion exchange separations. Stable in pH range of 2.5 to 8.
313,000.00 - ₹359,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific 30703-054630 APS-2 (Amino) Column - Length 50mm, 4.6mm ID, 3µm particle size 0188882 Mfr # 30703-054630
COMPARE
4.6 50 3
₹313,000.00INR / Each
Thermo Scientific 30703-154630 APS-2 (Amino) Column - Length 150mm, 4.6mm ID, 3µm particle size 0188890 Mfr # 30703-154630
COMPARE
4.6 150 3
₹359,000.00INR / Each
Thermo Scientific 30705-254630 APS-2 (Amino) Column - Length 250mm, 4.6mm ID, 5µm particle size 0188909 Mfr # 30705-254630
COMPARE
4.6 250 5
₹342,000.00INR / Each