Thermo Scientific Finnpipette Dispensers
Simple-to-use units offer consistent measuring results
  • Ceramic piston for dimensional stability and functional safety
  • Graduated gauge and digital-engaging volumetric adjustment provide absolute reproducibility
  • Ergonomic sure-grip operation
  • Highest degree of chemical and thermal resistance in medium-conveying components
These dispensers offer simple, rapid dispensing with reliable results through the unique combination of ceramic piston, graduated gauge, and digital volumetric adjustment device. This design provides absolute repoducibility and is calibrated for life. Dispensers are suitable for A32 thread.
225,000.00 - ₹319,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Division (mL) Max Dispense Volume (mL) Min Dispense Volume (mL)
Availability Pricing
Thermo Scientific 4421120 Finnpipette(R) Dispenser, 0.2 to 1.0 mL 0790315 Mfr # 4421120
COMPARE
0.05 1 0.2
₹225,000.00INR / Each
Thermo Scientific 4421140 Finnpipette(R) Dispenser, 1.0 to 5.0 mL 0790335 Mfr # 4421140
COMPARE
0.1 5 1
₹225,000.00INR / Each
Thermo Scientific 4421150 Finnpipette(R) Dispenser, 2.0 to 10.0 mL 0790345 Mfr # 4421150
COMPARE
0.25 10 2
₹225,000.00INR / Each
Thermo Scientific 4421160 Finnpipette(R) Dispenser, 5.0 to 30.0 mL 0790355 Mfr # 4421160
COMPARE
0.5 30 5
₹300,000.00INR / Each
Thermo Scientific 4421170 Finnpipette(R) Dispenser, 10.0 to 60.0 mL 0790365 Mfr # 4421170
COMPARE
1 60 10
₹319,000.00INR / Each