Thermo Scientific Digital Modular Dry Block Heaters
Precision temperature stability ensures reproducibility of results
  • Custom block modules provide flexibility for heating different size tubes
  • Built-in temperature sensing probe for outstanding temperature accuracy and control
  • Timer function allows the user to accurately monitor the heating time
These heaters have dynamic temperature stability providing outstanding temperature uniformity and stability. For a complete system order both a standard or advanced digital controled heater and the desired heating block modules.
Standard Digital Dry Block Heaters feature a proportional integrative derivative (PID) controller. PID control allows you to select a particular temperature with ease, enables rapid heat-up for samples, and provides excellent stability to maintain results. Temperatures can be set in 0.1°C increments. Other features include built-in temperature sensing probe, temperature offset function and timer.
200,000.00 - ₹265,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number Of Blocks Power (VAC)
Availability Pricing
Thermo Scientific 88870001 Digital Dry Block Heater, Single Block, 120V 3640306 Mfr # 88870001
COMPARE
1 120
₹225,000.00INR / Each
Thermo Scientific 88870002 Digital Dry Block Heater, Dual Block, 120V 3640307 Mfr # 88870002
COMPARE
2 120
₹213,000.00INR / Each
Thermo Scientific 88870003 Digital Dry Block Heater, Four Block, 120V 3640308 Mfr # 88870003
COMPARE
4 120
₹257,000.00INR / Each
Thermo Scientific 88870004 Digital Dry Block Heater, Single Block, 240V 3640309 Mfr # 88870004
COMPARE
1 240
₹200,000.00INR / Each
Thermo Scientific 88870005 Digital Dry Block Heater, Dual Block, 240V 3640310 Mfr # 88870005
COMPARE
2 240
₹221,000.00INR / Each
Thermo Scientific 88870006 Digital Dry Block Heater, Four Block, 240V 3640311 Mfr # 88870006
COMPARE
4 240
₹265,000.00INR / Each