Thermo Scientific Compact Digital Microplate Shaker
Precise and accurate shaking of up to four microplates
  • PID control for consistent shaking and precise temperature control
  • Digital display provides easy viewing
  • Compact design easily fits in incubators (non-CO2), chambers, and refrigerators (up to 60°C)
  • Use in continuous shaking or timer mode up to 99 hrs 59 min
  • Automatic overload shutdown
  • Standard configuration: 4 microplates (tube accessory sold separately)
294,000.00 - ₹303,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Load (lbs) Max Speed (rpm) Power (VAC)
Availability Pricing
Thermo Scientific Microplate Shaker, 100-240 VAC; US Plug 0472511 Mfr # 88882005
COMPARE
8.8 1200 100 to 240
₹294,000.00INR / Each
Thermo Scientific 88882006 Microplate Shaker, 100-240 VAC; International Plug 0472513 Mfr # 88882006
COMPARE
8.8 1200 100 to 240
₹303,000.00INR / Each