Thermo Scientific™ BioBasic™ AX Polyethyleneimine HPLC Columns
Stable polymeric coating is suitable for a wide range of analyte separations
  • Weak anion exchange for multiple charged analytes
  • Can be used for hydrophilic interaction chromatography (HILIC)
  • Proprietary, protein-friendly, ion exchange ligands of polyethyleneimine (PEI) bound to pure deactivated silica
  • Low hydrophobicity
Thermo Scientific™ BioBasic™ HPLC columns provide improved performance for biological samples including peptides, proteins, and other biomolecules. The high purity silica and tailored pore sizes provide superior performance in terms of improved resolution, efficiency, reproducibility, and column life.
BioBasic AX Columns utilize a polymeric secondary-tertiary amine (polyethyleneimine) ligand that is bound to a highly pure, base-deactivated silica that provides separation of proteins, peptides, other anionic species, and polar molecules. The polyethyleneimine bonded phase allows for additional chromatographic options, including hydrophilic interaction chromatography (HILIC) — ideal for separation of sugars and other highly polar species. BioBasic AX columns are i
356,000.00 - ₹398,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Phase
Availability Pricing
Thermo Scientific Biobasic 73105-052130 AX HPLC Column, Polyethyleneimine, 5 µm, 50 mm L x 2.1 mm ID 0190124 Mfr # 73105-052130
COMPARE
2.1 50 Polyethyleneimine (PEI)
₹356,000.00INR / Each
Thermo Scientific Biobasic 73105-152130 AX HPLC Column, Polyethyleneimine, 5 µm, 150 mm L x 2.1 mm ID 0190144 Mfr # 73105-152130
COMPARE
2.1 150 Polyethyleneimine (PEI)
₹398,000.00INR / Each