Thermo Scientific Barnstead MicroPure Water Purification Systems
18.2 MΩ-cm water in a compact system
  • Feed water monitoring for maximizing cartridge life
  • Variable flow control knob for dispensing
  • Quick-change cartridges for rapid cartridge change, even mid-operation
  • Optional integrated 6-L feed water reservoir for areas without direct access to a water line
Barnstead MicroPure systems deliver up to 15 L/day of Type I ultrapure water from a pretreated feed water supply. Water product quality exceeds international standards ASTM Type I, ISO 3696 Grade 1, ASTM D1193 and CLSI-CLRW.
These compact systems are an economical option for a wide range of applications needs from basic buffer preparations to highly sensitive protocols such as HPLC, PCR, or applications that require precise TOC removal and measurement.
The ergonomically designed controller is backlit and tilts for easy operation and better visibility. Dispenser allows drop by drop or quick dispensing.
Recirculation pump prevents bacterial growth during standstill and allows for constant monitoring of all system functions, allowing for the immediate display of fault messages if a problem is detected.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
ASTM Water Grade With Ultrafilter With UV Lamp
Availability Pricing
Thermo Scientific Barnstead 50132373 MicroPure water system, UV 9927130 Mfr # 50132373
COMPARE
Type I Yes No
Call For Price
Thermo Scientific Barnstead 50132374 MicroPure UV Reservoir Feed 9927131 Mfr # 50132374
COMPARE
Type I Yes No
Call For Price
Thermo Scientific Barnstead 50132372PR MicroPure UV/UF Reservoir Feed, complete package 9927141 Mfr # 50132372
COMPARE
Type I Yes Yes