Thermo Scientific Advanced Digital Modular Dry Block Heaters with Touch Screen
Precision temperature stability ensures reproducibility of results
  • Custom block modules provide flexibility for heating different size tubes
  • Built-in temperature sensing probe for outstanding temperature accuracy and control
  • Timer function allows the user to accurately monitor the heating time
These heaters have dynamic temperature stability providing outstanding temperature uniformity and stability. For a complete system order both a standard or advanced digital controled heater and the desired heating block modules.
Advanced Digital Dry Block Heaters feature a proportional integrative derivative (PID) controller. PID control allows you to select a particular temperature with ease, enables rapid heat-up for samples, and provides excellent stability to maintain results. Temperatures can be set in 0.1°C increments. Heaters also have an easy-to-use touch screen interface and built-in over-temperature protection. The advanced model also has 10 user-settable programs; each program supports up to 5 sequential steps of different temperatures and duration times. The optional external temperature sensor allows for direct temperature monitoring within the block. When the external temperature sensor is connected, the internal temperature sensor will disconnect automatically. Other features include built-in temperature sensing probe, temperature offset function and timer.
321,000.00 - ₹401,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number Of Blocks Power (VAC) Description
Availability Pricing
Thermo Scientific 88870007 Advanced Digital Dry Block Heater, Touch Screen, Single Block 120V 3640300 Mfr # 88870007
COMPARE
1 120 Advanced Digital Dry Block Heater, Touch Screen, Single Block 120V
₹328,000.00INR / Each
Thermo Scientific 88870008 Advanced Digital Dry Block Heater, Touch Screen, Dual Block, 120V 3640301 Mfr # 88870008
COMPARE
2 120 Advanced Digital Dry Block Heater, Touch Screen, Dual Block, 120V
₹321,000.00INR / Each
Thermo Scientific 88870012 Advanced Digital Dry Block Heater, Touch Screen, Four Block, 240V 3640305 Mfr # 88870012
COMPARE
4 240 Advanced Digital Dry Block Heater, Touch Screen, Four Block, 240V
₹401,000.00INR / Each