Texwipe Cleanroom LDPE Adhesive Tapes
Ideal for sealing and packaging materials in cleanroom
  • Packaged in press-and-seal cleanroom bags
  • Individually lot coded for ease of traceability
  • Double bagged for use in cleanroom environments
These adhesive tapes are ideal for color coding and product identification for most applications and environments. They are well suited for sealing compound surfaces and packaging materials within your cleanroom. Constructed of low-density polyethylene (LDPE) backing the tapes feature strong adhesion and are easy to tear by hand.
The TPA series tapes feature an acrylic adhesive, while the TPR Series tape features a rubber adhesive.
224,000.00 - ₹270,000.00INR / Case of 48
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Material Width (in)
Availability Pricing
Texwipe TPA LDPE/Acrylic Clear Cleanroom Adhesive Tape, 1" W x 108' L; 48/CS 3367650 Mfr # TPA1048CL
COMPARE
Clear LDPE with acrylic adhesive 1
₹224,000.00INR / Case of 48
Texwipe TPA LDPE/Acrylic Blue Cleanroom Adhesive Tape, 1" W x 108' L; 48/CS 3367651 Mfr # TPA1048BL
COMPARE
Blue LDPE with acrylic adhesive 1
₹237,000.00INR / Case of 48
Texwipe TPA LDPE/Acrylic White Cleanroom Adhesive Tape, 1" W x 108' L; 48/CS 3367652 Mfr # TPA1048WH
COMPARE
White LDPE with acrylic adhesive 1
₹224,000.00INR / Case of 48
Texwipe TPA LDPE/Acrylic White Cleanroom Adhesive Tape, 2" W x 108' L; 24/CS 3367653 Mfr # TPA2024WH
COMPARE
White LDPE with acrylic adhesive 2
₹229,000.00INR / Case of 24
Texwipe TPA LDPE/Acrylic White Cleanroom Adhesive Tape, Perforated, 2" W x 108' L; 24/CS 3367654 Mfr # TPA2036WHC
COMPARE
White, perforated LDPE with acrylic adhesive 2
₹270,000.00INR / Case of 24
Texwipe TPR LDPE/Rubber Blue Cleanroom Adhesive Tape, 1" W x 108' L; 48/CS 3367655 Mfr # TPR1048BL
COMPARE
Blue LDPE with rubber adhesive 1
₹260,000.00INR / Case of 48