Testo Pocket-Line Thermoanemometer
Ergonomically designed air velocity and temperature meter in one
  • Small, handy and easy to operate
This rugged meter is pocket sized, so it can always be with you when you need it. Ideal for measuring general airflow or when monitoring more specific HVAC conditions. Professional-grade sensors produce accuracies is uncommon in a meter this size. The menu-driven three button design allows for easy selection of the meters function. Both models feature air velocity, ambient temperature, min/max/hold functions, selectable velocity and temperature units, and a large easy-to-read backlit display. Unique hard shell protective cap snaps on and off the meter. When in use, the cap fully encases the meter, protecting everything including the display screen and sensors.
57,600.00 - ₹93,500.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Air Velocity Accuracy Air Velocity Resolution Max Air Velocity (ft/min)
Availability Pricing
Testo 0560 4101 Pocket Line Thermoanemeter, 1.5" vane 1032351 Mfr # 0560 4101
COMPARE
±(2% of reading + 40 ft/min) 1 ft/min (0.01 m/s) 3937
₹57,600.00INR / Each
Testo 0560 4102 Pocket Line Thermoanemeter with Humidity 1032352 Mfr # 0560 4102
COMPARE
±(2% of reading + 40 ft/min) 1 ft/min (0.01 m/s) 3937
₹93,500.00INR / Each