Testo Humidity / Dew Point Monitoring Transmitters
Ideal for continuous monitoring of dryers, furnaces, and compressed air systems
  • IP65 rated housing protects in heavy-duty environments
  • Pressure tight up to 735 psig (50 bar)
Transmitters are powered by 24 VDC and have an output of 4 to 20 mA. The capacitive sensor is accurate, rugged, and offers long-term stability with temperature resistance. The IP65 rated housing protects the unit from dust, sprays, and electromagnetic interference. A unique rotating body allows optimal positioning, while the three-button keypad makes setup and adjustments fast and easy. These units can be programmed directly via a computer. The process connection is G 1/2" stainless steel screw-in thread or 1/2" NPT that easily threads into existing sample ports. Select from two styles of transmitters with or without a display — for your demanding applications. Models with a display are more commonly used for monitoring at the application level, and features Min/Max, selectable display °F/°C display, Ftd or Ctd (temperature dew point), Atd (atmospheric dew point °F or °C), %RH, ppm, and mg/m3. Models without a display unit are ideal for OEM applications where monitoring is done in a process setting.
The optional quick-connect measuring chamber 37463-52 has a standard G 1/2 connection and ensures the transmitter has the optimal flow past the sensor. Use the optional two-output signal switch 37463-54 to add alarm capability when the selected limit value is exceeded.
574,000.00 - ₹726,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Display Type Max Dew Point Range (° C) Min Dew Point Range (° F)
Availability Pricing
Testo 0555 6742 Transmitter, Dew Point, 1/2" NPT Connection 3746304 Mfr # 0555 6742
COMPARE
None 30 -76
₹574,000.00INR / Each
Testo 0555 6743 Transmitter, Dew Point, with Display, G 1/2" Connection 3746306 Mfr # 0555 6743
COMPARE
LCD 30 -76
₹726,000.00INR / Each
Testo 0555 6744 Transmitter, Dew Point, with Display, 1/2" NPT Connection 3746308 Mfr # 0555 6744
COMPARE
LCD 30 -76
₹726,000.00INR / Each