Tegam Intirinsically Safe Thermocouple Thermometers
ATEX-compliant intrinsically safe thermometers—ideal for use in hazardous locations
  • Extra-long battery life—lasts about 2000 hours using three AA batteries!
  • ±(0.04% of reading + 0.3°C) accuracy
  • Accepts thermocouple types J, K, T, E, N, R, S, and B
  • Ergonomically designed to minimize user fatigue
  • Easy-to-clean seamless case
Ensure worker safety when measuring temperatures in potentially hazardous locations. These low energy intrinsically safe thermometers safeguard against ignition when used in environments where flammable liquids, gases, vapors, and dust are present. Both 921B and 922B models accept type J, K, T, E, N, R, S, and B thermocouple probes and have accuracy of ±(0.04% of reading + 0.3°C). Use the probe offset feature to zero out temperature probe errors, significantly improving overall measurement uncertainty. Thermometers provide exceptionally long battery life of 2000 hours using three AA batteries.
Increase productivity, efficiency, and quality using real-time statistical data. Thermometers display min, max, average, range, and standard deviation on demand, providing critical information and helping to analyze manufacturing process to make immediate adjustment for maximum efficiency and quality. Press the HOLD button to freeze display for reading later.
Easily view the readings on a large backlit LCD. The ergonomically designed thermometer allows for one-handed operation and minimizes user fatigue during extended use. The smooth, seamless case resists dirt and dust and is easy to clean. Available in single- or dual-input model.
188,000.00 - ₹221,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Number Of Probe Channels Types Of Probes Accepted
Availability Pricing
Tegam 921B Intrinsically Safe Thermocouple Thermometer, Single-input 7890932 Mfr # 921B
COMPARE
1820 1 Types J, Type K, Type T, Type E, Type B, Type N, Type R, Type S
₹188,000.00INR / Each
Tegam 922B Intrinsically Safe Thermocouple Thermometer, Dual-input 7890933 Mfr # 922B
COMPARE
1820 2 Types J, Type K, Type T, Type E, Type B, Type N, Type R, Type S
₹221,000.00INR / Each