Tegam Digital Thermocouple Thermometers
Superior accuracy and long battery life for uninterrupted, dependable temperature measurements
  • Extra-long battery life—lasts about 2000 hours using three AA batteries!
  • ±(0.04% of reading + 0.3°C) accuracy
  • Accepts thermocouple types J, K, T, and E
  • Ergonomically designed to minimize user fatigue
  • Easy-to-clean seamless case
Get the accuracy and dependability you need to measure temperature using the 911B and 912B thermocouple thermometers. Both models accept type J, K, T, and E thermocouple probes and have accuracy of ±(0.04% of reading + 0.3°C). Use the probe offset feature to zero out temperature probe errors, significantly improving overall measurement uncertainty. Thermometers provide exceptionally long battery life of 2000 hours using three AA batteries.
Increase productivity, efficiency, and quality using real-time statistical data. These low-cost models display min, max, average, range, and standard deviation on demand, providing critical information and helping to analyze manufacturing process to make immediate adjustment for maximum efficiency and quality. Press the HOLD button to freeze display for reading later.
Easily view the readings on a large backlit LCD. The ergonomically designed thermometer allows for one-handed operation and minimizes user fatigue during extended use. The smooth, seamless case resists dirt and dust and is easy to clean. Available in single- or dual-input model.
124,000.00 - ₹151,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Number Of Probe Channels Types Of Probes Accepted
Availability Pricing
Tegam 911B Digital Thermocouple Thermometer, Single Input 7890930 Mfr # 911B
COMPARE
1372 1 Types J, Type K, Type T, Type E
₹151,000.00INR / Each
Tegam 912B Digital Thermocouple Thermometer, Dual input 7890931 Mfr # 912B
COMPARE
1372 2 Types J, Type K, Type T, Type E
₹124,000.00INR / Each