Swift Optical M3-F Series Forensic Microscope
Compare images in both micro and macro environments
  • View side-by-side, full left and right, or overlapping images
  • Rechargeable LED lights keep power cord out of the way
  • A 45° inclined Siedentopf head provides user comfort
  • Smooth, ball-bearing mounted quad objectives for precise positioning of objectives
  • Attach an imaging device to the trinocular port
The M3-F trinocular forensic microscope offers highly professional features at a competitive price. Easily view images fully from the left microscope or right microscope or compare images from two M3 microscopes by putting them side-by-side or by overlapping. A comparison bridge acts as a director of image paths from the two M3 microscopes.
Microscope comes with widefield 10x/18 mm eyepieces and DIN standard 4x (0.10 N.A.), 10x (0.25 N.A.), 40xR (0.65 N.A.) achromat objectives and 1x macro objective. Use the macro lens with a large working distance to view larger, bulkier items. Place your sample for examination on a gliding stage plate, black and white contrast plate, or specimen cup depending on your application.
Top and bottom cordless LED illumination provide energy efficient white light. Lights last up to 25,000 hours per charge so there is no need to deal with power cords while working; charging turns off automatically when battery is fully charged.
582,000.00 - ₹840,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Output Size Description
Availability Pricing
Swift Optical M3-F Forensic Comparison Trinocular Microscope 4892678 Mfr # M3-F
COMPARE
None No Display Forensic Comparison Trinocular Microscope
₹582,000.00INR / Each
Swift Optical M3-F-BTI1 Forensic Comparison Microscope with Detachable 8" Tablet 4892679 Mfr # M3-F-BTI1
COMPARE
WiFi, mini HDMI 8" Tablet Forensic Comparison Microscope with Detachable 8" Tablet
₹774,000.00INR / Each
Swift Optical M3-F-BTI2 Forensic Comparison Microscope with Detachable 10" Tablet 4892680 Mfr # M3-F-BTI2
COMPARE
WiFi, mini HDMI 10" Tablet Forensic Comparison Microscope with Detachable 10" Tablet
₹840,000.00INR / Each