Swift Optical M3-B Micro/Macro Cordless Field Microscopes
Cordless microscope uses rechargeable batteries for easy field use
  • Versatile design allows for use as a microscope, macroscope, or in the field
  • Sturdy detachable tripod legs fold for easy storage and sturdy even on rugged outdoor surfaces
  • Energy efficient, rechargeable LED illumination allows up to 40 hours of use on a single 8 hour charge
Rechargeable batteries, tripod legs, and a carrying handle make this microscope ideal for field use — take it wherever you need it! Durable all-metal construction withstands outdoor environments. Versatile microscope features three objective lenses for microscopic use and one lens for macroscopic viewing of larger objects. Top and bottom LED illumination uses little power allowing up to 40 hours of field use before recharging is necessary. Stage position adjusts for micro- and macro-viewing. Tripod legs fold for easy storage; for added versatility, microscope can be mounted on a standard camera tripod for easier field use.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Head Configuration
Availability Pricing
Swift Optical M3-B Micro/Macro Cordless Field Microscope, Binocular 0310026 Mfr # M3-B
COMPARE
Binocular