SUPERLOK SRU Reducing Union Fittings, Stainless Steel
Achieve leak-free, high-performance connections with SUPERLOK Tube Fittings, designed for reliability in even the most demanding applications
  • Double and single ferrule compression fittings for secure connections
  • Easy installation with minimal steps for fast assembly
  • High-quality stainless steel construction
  • Wide range of fitting types for versatile industrial applications
  • Gauge-able fittings ensure proper tightening and safety
  • Reducing unions connect tubing of two different sizes, from a larger to a smaller flow path
SUPERLOK Tube Fittings are engineered to provide the most reliable connections in industries where safety and performance are critical. These products are easy to install, durable, and resistant to leaks. Made from top-grade stainless steel, SUPERLOK fittings are ideal for high-pressure systems, gas applications, and more. Ensure secure installation with precise gaugeability and superior construction.
2,900.00 - ₹6,200.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Compression Size / Tube Od (Port 2) (in) Flow Pattern
Availability Pricing
Superlok USA Reducing Union, 1/8" X 1/16", Stainless Steel; 1/Each 6780165 Mfr # SRU2-1-S316
COMPARE
1/8 1/16 Straight through
₹3,900.00INR / Each
Superlok USA Reducing Union, 1/4" X 1/8", Stainless Steel; 1/Each 6780166 Mfr # SRU4-2-S316**
COMPARE
1/4 1/8 Straight through
₹2,900.00INR / Each
Superlok USA Reducing Union, 3/8" X 1/4", Stainless Steel; 1/Each 6780167 Mfr # SRU6-4-S316
COMPARE
3/8 1/4 Straight through
₹3,900.00INR / Each
Superlok USA Reducing Union, 1/2" X 1/4", Stainless Steel; 1/Each 6780168 Mfr # SRU8-4-S316
COMPARE
1/2 1/4 Straight through
₹6,200.00INR / Each