SUPERLOK SMC Male Connector Fittings, Stainless Steel
Achieve leak-free, high-performance connections with SUPERLOK Tube Fittings, designed for reliability in even the most demanding applications
  • Double and single ferrule compression fittings for secure connections
  • Easy installation with minimal steps for fast assembly
  • High-quality stainless steel construction
  • Wide range of fitting types for versatile industrial applications
  • Gauge-able fittings ensure proper tightening and safety
  • Connects tubing to a female-threaded component with male NPT threads
SUPERLOK Tube Fittings are engineered to provide the most reliable connections in industries where safety and performance are critical. These products are easy to install, durable, and resistant to leaks. Made from top-grade stainless steel, SUPERLOK fittings are ideal for high-pressure systems, gas applications, and more. Ensure secure installation with precise gaugeability and superior construction.
2,100.00 - ₹4,300.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Flow Pattern Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
Superlok USA Male Connector, 1/16" O.D. X 1/8" NPT(M),Stainless Steel; 1/Each 6780169 Mfr # SMC1-2N-S316
COMPARE
1/16 Straight through 1/8
₹3,100.00INR / Each
Superlok USA Male Connector, 1/8" O.D. X 1/8" NPT(M),Stainless Steel; 1/Each 6780170 Mfr # SMC2-2N-S316
COMPARE
1/8 Straight through 1/8
₹2,400.00INR / Each
Superlok USA Male Connector, 1/4" O.D. X 1/8" NPT(M),Stainless Steel; 1/Each 6780171 Mfr # SMC4-2N-S316**
COMPARE
1/4 Straight through 1/8
₹2,200.00INR / Each
Superlok USA Male Connector, 1/4" O.D. X 1/4" NPT(M), Stainless Steel; 1/Each 6780172 Mfr # SMC4-4N-S316**
COMPARE
1/4 Straight through 1/4
₹2,100.00INR / Each
Superlok USA Male Connector, 3/8" O.D. X 1/4" NPT(M),Stainless Steel; 1/Each 6780173 Mfr # SMC6-4N-S316**
COMPARE
3/8 Straight through 1/4
₹2,600.00INR / Each
Superlok USA Male Connector, 3/8" O.D. X 3/8" NPT(M),Stainless Steel; 1/Each 6780174 Mfr # SMC6-6N-S316
COMPARE
3/8 Straight through 3/8
₹2,900.00INR / Each
Superlok USA Male Connector, 1/2" O.D. X 3/8" NPT(M), Stainless Steel; 1/Each 6780175 Mfr # SMC8-6N-S316
COMPARE
1/2 Straight through 3/8
₹3,900.00INR / Each
Superlok USA Male Connector, 1/2" O.D. X 1/2" NPT(M),Stainless Steel; 1/Each 6780176 Mfr # SMC8-8N-S316
COMPARE
1/2 Straight through 1/2
₹4,100.00INR / Each
Superlok USA Male Connector, 3/8" O.D. X 3/8" NPT(M),Stainless Steel; 1/Each 6780183 Mfr # SMC6-6N-S316
COMPARE
3/8 Straight through 3/8
₹2,900.00INR / Each
Superlok USA Male Connector, 1/2" O.D. X 1/4" NPT(M), Stainless Steel; 1/Each 6780184 Mfr # SMC8-4N-S316
COMPARE
1/2 Straight through 1/4
₹3,800.00INR / Each
Superlok USA Male Connector, 1/2" O.D. X 1/2" NPT(M),Stainless Steel; 1/Each 6780185 Mfr # SMC8-8N-S316
COMPARE
1/2 Straight through 1/2
₹4,300.00INR / Each
Showing 1- 11 of 11