Submersible pH Electrodes
Ideal for field applications
  • Completely submersible up to the last one foot prior to the meter connection
  • Extended cable lengths up to 100 feet
These ABS plastic electrodes are designed for field applications. Annular junction provides fast response and resists pressure effects of submersion. BNC connection is compatible with most pH meters. 100 foot cable length electrode includes preamplification.
30,800.00 - ₹41,500.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Electrode pH submersible 25 ft cable length BNC
₹30,800.00INR / Each
COMPARE
Electrode pH submersible 50 ft cable length BNC
₹41,500.00INR / Each