Stuart® Vortex Mixers
Compact unit perfect for individual work stations
  • Mix a wide variety of tube sizes and shapes with the included rubber cup head
  • Heavy die-cast body won't dance across your bench
  • Use for continuous mixing — housing design supports use with a lab rod
Vortex mixer features a robust die-cast body to prevent traveling while on, and has an integrated port to support a lab rod for holding vessels in place during continuous operation.
30,100.00 - ₹39,100.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Power (VAC) Description
Availability Pricing
COMPARE
2500 100 to 230 Fixed-Speed
₹37,000.00INR / Each
COMPARE
2500 120 / 230 Variable Speed
₹39,100.00INR / Each
COMPARE
4500 100 to 240 Variable Speed Mini Vortex Mixer
₹30,100.00INR / Each