Cole-Parmer® RS-200 Series Roller Shakers
Gentle rocking and rolling action for complete mixing
  • Safe for use in cold rooms down to 39 °F (4 °C) or in incubators to 104 °F (40 °C), up to 80% humidity
  • Continuous and quiet operation
  • Side panels ensure sample bottles stay on the shaker
Solidly constructed roller shakers are ideal for mixing blood samples, viscous substances, and liquid-solid suspensions where minimum aeration is required. Select shakers with six or nine tube rollers to mix multiple samples at the same time while saving benchtop space. Order the stacking system 04653-09 separately to stack up to three shakers to meet high throughput applications without taking valuable bench space.
141,000.00 - ₹334,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Power (VAC) Width (in)
Availability Pricing
COMPARE
33 230 10.23
₹141,000.00INR / Each
COMPARE
33 120 10.23
₹232,000.00INR / Each
COMPARE
33 230 14.96
₹162,000.00INR / Each
COMPARE
33 120 14.96
₹292,000.00INR / Each
COMPARE
60 230 10.23
₹158,000.00INR / Each
COMPARE
60 120 10.23
₹286,000.00INR / Each
COMPARE
60 230 14.96
₹188,000.00INR / Each
COMPARE
60 120 14.96
₹334,000.00INR / Each
Showing 1- 8 of 8