Cole-Parmer® ST-200 and SHP-200 Series 3-Position Stirrer
Three independent stirring or heating/stirring positions yet only measures 23-5/8"L x 10-5/8"W (60 x 27 cm)
  • Powerful magnets for strong magnetic coupling and stirring capacity
  • Easily accommodate a 2-L beaker in each position
Three independently controlled position units only require one power source. Run three separate tests at the same time or run multiple samples under the same conditions.
91,200.00 - ₹139,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Three Position Magnetic Stirrer; 230 VAC, 50 Hz
₹91,200.00INR / Each
COMPARE
Three Position Stirring Hot Plate; 230 VAC, 50 Hz
₹139,000.00INR / Each