Storage Cabinets

Stack these storage cabinets or mount them on a wall. Cabinet frame is made of heavy-duty welded steel with a baked-enamel finish. Steel shelves and center posts support tough, removable drawers. Front label holders let you identify contents quickly. Cabinets measure 17"W x 11"H x 11"D (43 x 28 x 28 cm).
54,600.00 - ₹69,600.00INR / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Number of Bins Width (in)
Availability Pricing
Akro-Mils 19-320 Storage Cabinet, 20-Drawer; 17" x 11" x 11" 0978601 Mfr # 19-320
COMPARE
11 20 17
₹69,600.00INR / Each
Akro-Mils 19-416 Storage Cabinet, 16-Drawer; 17" x 11" x 11" 0978602 Mfr # 19-416
COMPARE
11 16 17
₹60,300.00INR / Each
Akro-Mils 19-715 Storage Cabinet, 15-Drawer; 17" x 11" x 11" 0978603 Mfr # 19-715
COMPARE
11 15 17
₹69,100.00INR / Each
Akro-Mils 19-909 Storage Cabinet, 9-Drawer; 17" x 11" x 11" 0978604 Mfr # 19-909
COMPARE
11 9 17
₹54,600.00INR / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat