SteriMax Electric Sterilizing Tube
Sterilize loops, needles and instruments without flames
  • Use in laboratories and safety cabinets where gases and flames are not permitted
  • Sterilize loops, needles, and instruments within seconds
  • Durable stainless steel construction withstands harsh environments and cleans easily
Durable stainless steel construction withstands harsh environments and temperatures The unique double tube system, which is made of robust quartz glass and wear- resistant ceramic, reaches optimal temperature of 900°C (1650°F) within minutes. Simply insert your instrument into the tube for fast, effective sterilization. An integrated spray protector provides additional safety when handling pathogenic material. The stainless steel design withstands the most extreme laboratory conditions and is easy to clean. Low-profile, stable housing allows ergonomic operation.
Use the built-in flat tray to conveniently rest your loops.
152,000.00 - ₹171,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Chamber Depth (in) Chamber Diameter (in) Power (VAC)
Availability Pricing
SteriMax 5.002.000 Electrical Sterilizer, 118 mm L X 15 mm Dia, 120 VAC WW-36302-15 Mfr # 5.002.000
COMPARE
4.625 0.625 115
₹152,000.00INR / Each
SteriMax 5.001.000 Electrical Sterilizer, 118 mm L X 15 mm Dia, 230 VAC WW-36302-16 Mfr # 5.001.000
COMPARE
4.625 0.625 230
₹156,000.00INR / Each
SteriMax 5.102.000 Electrical Sterilizer, 161 mm L X 21 mm Dia, 120 VAC WW-36302-17 Mfr # 5.102.000
COMPARE
6.375 0.875 115
₹171,000.00INR / Each