Stainless Steel Cell Spreaders
Spread bacterial cultures evenly
  • Constructed of durable and reusable stainless steel
  • Apply cultures over an entire petri dish or plate with the triangular-shaped spreader
  • Autoclave or flame sterilize
Stainless steel spreaders have a smooth triangular end with straight spreading bar to reduce the risk of damaging the agar. Both small and large sizes are 302 mm (8") long.
9,100.00 - ₹9,600.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Material Width (mm) Description
Availability Pricing
COMPARE
Stainless Steel 16 Cell Spreader, Stainless Steel, 16-mm Spreading Bar
₹9,600.00INR / Each
COMPARE
Stainless Steel 25 Cell Spreader, Stainless Steel, 25-mm Spreading Bar
₹9,100.00INR / Each