Spill Kit Refills
Arrest spills anytime-anywhere
  • No need to purchase new wheeled bins or drums
  • Effectively prevent and control possible liquid splashes, spills, or
  • pollution of spilled liquids
  • Keep the storage environment of oily and chemical drums clean
  • Comply with EPA, SPCC, and NPDES regulations to keep you safe
Spill control products are essential to any well-organized lab. When
spills happen in the workplace, the type of spill needs to be quickly
identified and the correct products used for spill containment
and cleanup. Universal (gray): oils, coolant, solvents, and water.
Chemicals (yellow): acids, corrosives, and unknown hazardous
chemicals. Oil only (white): oils, fuels, and other oil-based liquids;
not water.
Melt-blow polypropylene (MBPP) absorbent materials feature high
absorption rate, rapid absorption, good cleaning effect; don’t react
with polar, toxic, hazardous chemicals; and are safe to use, causing
no harm to personal health.
6,900.00 - ₹7,900.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Chemical Treated Length (yd) Width (in)
Availability Pricing
SYSBEL SKS001G Spill Kit Refill, Universal; 10 gal.(38 L) WW-15985-90 Mfr # SKS001G
COMPARE
₹7,500.00INR / Each
SYSBEL SKS001Y Spill Kit Refill, Chemicals; 10 gal.(38 L) WW-15985-91 Mfr # SKS001Y
COMPARE
₹7,900.00INR / Each
SYSBEL SKS001W Spill Kit Refill, Oil only; 10 gal.(38 L) WW-15985-92 Mfr # SKS001W
COMPARE
₹6,900.00INR / Each