Speirs Robertson AMTR and AMTS Series Laboratory Work Tables
High quality tables meeting cleanroom and laboratory requirements
  • Sturdy tables for laboratory use with Trepsa or Stanless Steel worktops
The laboratory tables are made to the highest standard with Trespa or Stainless worktops supported by rigid frames with levelling feet. Frames are made from strong steel box section welded together and epoxy powder coated white. Accessories such as shelves and monitor supports are available. They are easy to install and require no maintenance.
691,000.00 - ₹784,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Depth (in) Height (in) Width (in)
Availability Pricing
Speirs Robertson AMTS-75x75 AMTS Stainless Steel (Top) Laboratory Work Table 75x75 cm 1150051 Mfr # AMTS-75X75
COMPARE
30 30 30
₹691,000.00INR / Each
Speirs Robertson AMTS-90x75 AMTS Stainless Steel (Top) Laboratory Work Table 90x75 cm 1150052 Mfr # AMTS-90X75
COMPARE
30 30 36
₹691,000.00INR / Each
Speirs Robertson AMTR-75x75 AMTR White Trespa (Top) Laboratory Work Table 75x75 cm 1150054 Mfr # AMTR-75X75
COMPARE
30 30 30
₹784,000.00INR / Each