Speirs Robertson AMD-SR Series Instrument Isolation Tables
Improves performance of sensitive instruments by shielding them from vibrations
  • Improve instrument performance by removing vibrations
  • High performance isolators available in a choice of performance levels
  • Suitable for instruments from 10kg up to 200kgs
Save time and money by making sure your expensive high performance instruments are not underperforming due to a poor working environment. Vibrations limit the performance of sensitive equipment and manufacturer’s specifications usually state that performance depends on working in the right environment. The tables remove these vibrations allowing you to get the performance you expect. Active tables require a supply of clean dry compressed air of at least 5Bar (10 Bar max) via a 6mm OD airline. Platforms are simple to install, easy to use and maintenance free.
912,000.00 - ₹1,426,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Description
Availability Pricing
Speirs Robertson AMD-SR-75X75-SS AMD-SR Instrument Table, Sorbothane Isolators; 75x75 cm 1150033 Mfr # AMD-SR-75X75-SS
COMPARE
200 440 AMD-SR Instrument Table, Sorbothane Isolators; 75x75 cm
₹912,000.00INR / Each
Speirs Robertson AMD-SR-90X75-SS AMD-SR Instrument Table, Sorbothane Isolators; 90x75 cm 1150034 Mfr # AMD-SR-90X75-SS
COMPARE
200 440 AMD-SR Instrument Table, Sorbothane Isolators; 90x75 cm
₹1,005,000.00INR / Each
Speirs Robertson AMD-SR-120X75-SS AMD-SR Instrument Table, Sorbothane Isolators; 120x75 cm 1150035 Mfr # AMD-SR-120X75-SS
COMPARE
200 440 AMD-SR Instrument Table, Sorbothane Isolators; 120x75 cm
₹1,426,000.00INR / Each