Speirs Robertson AMD-SB Series Stainless Steel Balance Isolation Tables
Improves performance of balances
  • Pharma grade stainless worktops meet stringent cleanliness requirements
  • Balance performance maximised by elimination of vibrations
  • Custom designed elastomeric isolators built into worktop
  • Ideal for larger balances, thermal analysers and similar instruments
Creates the best environment for your balance and by optimising user comfort improves both accuracy and efficiency of measurements. The range of balance tables have been carefully designed to remove vibrations which limit balance performance and provide a tailored working environment for the user. The custom designed Sorobthane isolators are extremely efficient in removing vibrations. The stainless worktops meet stringent lab cleanliness requirements. Options include acoustic hoods, armrest bars, shelves and monitor stands, allowing you to configure a table to suit you specific needs. Table are simple to install and require no maintenance.
752,000.00 - ₹1,059,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Height (in) Width (in) Capacity (lbs)
Availability Pricing
Speirs Robertson AMD-SB-75x75 AMD-SB Balance Table, Sorbothane Isolators; 75x75 cm 1150026 Mfr # AMD-SB-75X75
COMPARE
30 30 55
₹851,000.00INR / Each
Speirs Robertson AMD-SB-90x75 AMD-SB Balance Table, Sorbothane Isolators; 90x75 cm 1150027 Mfr # AMD-SB-90X75
COMPARE
30 36 55
₹752,000.00INR / Each
Speirs Robertson AMD-SB-120x75 AMD-SB Balance Table, Sorbothane Isolators; 120x75 cm 1150028 Mfr # AMD-SB-120X75
COMPARE
30 48 55
₹1,059,000.00INR / Each