Spectra/Por 3 Regenerated Cellulose (RC) Dialysis Tubing
Get gentle separation and purification of solutes
  • Available in a variety of diameters and flat widths
  • Max MWCO 3,500 daltons
The RC Dialysis tubing is packaged dry to provide you with easy handling and storage. Available in 15 meter rolls to provide better economy for applications requiring larger amounts of tubing.
Most of the dry tubing rolls contain glycerin as a humectant which is easily removed by soaking 30 min in water.
81,700.00 - ₹87,000.00INR / Box of 1
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Flat Width (mm) Max Molecular Weight Cut Off (Mwco) (Dalton)
Availability Pricing
Spectra Por 3 RC Dialysis Tubing, 3.5 kD MWCO, 45mm Flat Width, 28.6mm dia; 50' L 0290029 Mfr # 132724
COMPARE
28.6 45 3500
₹81,700.00INR / Box of 1
Spectra Por 3 RC Dialysis Tubing, 3.5 kD MWCO, 18mm Flat Width, 11.5mm dia; 50' L 0290030 Mfr # 132720
COMPARE
11.5 18 3500
₹81,700.00INR / Box of 1
Spectra Por 3 RC Dialysis Tubing, 3.5 kD MWCO, 54mm Flat Width, 34mm dia; 50' L 0290031 Mfr # 132725
COMPARE
34 54 3500
₹87,000.00INR / Box of 1