Socorex Calibrex Low-Volume Dispensers (with cover)
Precalibrated fixed stops ensure accurate dispensing
  • PFA-coated glass plunger prevents freezing when dispensing causticsolutions
  • Autoclavable
These bottletop dispensers are so easy to use! Simply rotate the handpiece and receiver to choose volume desired; raise the handpiece fully and depress to dispense. The low-volume models include a cover to protect the glass column. All models feature a sturdy delivery jet made of PTFE/ETFE. Dispensers fit bottles with 33-mm threaded screw tops and include adapters for 28-, 38-, and 45-mm bottles. Order bottles separately.
141,000.00 - ₹267,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Division (mL) Max Dispense Volume (mL) Min Dispense Volume (mL)
Availability Pricing
Socorex 844002 Low-volume dispenser, w/cover, 1-5 mL 0791445 Mfr # 844002
COMPARE
0.1 5 1
₹141,000.00INR / Each
Socorex 844004 Low-volume dispenser, w/cover, 1-10 mL - +/-0.7 0791450 Mfr # 844004
COMPARE
0.25 10 1
₹267,000.00INR / Each