Socorex Acura Single-Channel Adjustable-Volume Pipettors
Adjustable tip ejector for tip source flexibility
  • Fast easy calibration reduces pipettor downtime
  • Fully autoclavable to 248°F (121°C) — fully assembled
  • 3-year warranty on parts and labor including 2 years on the original calibration
  • Confirm your volume easily while in use
These pipettors combine numerous features and accuracy in a convenient package. Read the volume display without changing your hand position, ensuring the correct liquid volume is dispensed. The JustTip™ system enables easy adjustment of the tip ejector so that it can be used with a variety of tips. The tip ejector is positioned for easy use by either hand, even when wearing gloves.
Perform calibration quickly and easily, reducing pipettor downtime. Swift-set calibration requires no additional tools beyond a balance and distilled water. Pipettors are fully autoclavable to 248°F (121°C) fully assembled, reducing the time to bring a pipettor back to service after contamination.
154,000.00 - ₹229,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Min Volume (µL)
Availability Pricing
Socorex Acura Adjustable-Volume Pipettor, 2-20 µL; Each 2501506 Mfr # W810306
COMPARE
20 2
₹229,000.00INR / Each
Socorex Acura Adjustable-Volume Pipettor, 20-200 µL; Each 2501514 Mfr # W810312
COMPARE
200 20
₹229,000.00INR / Each
Socorex Acura Adjustable-Volume Pipettor, 100-1000 µL; Each 2501516 Mfr # W810314
COMPARE
1000 100
₹229,000.00INR / Each
Socorex Acura Adjustable-Volume Pipettor, 0.5-5mL; Each 2501518 Mfr # W810316
COMPARE
5000 500
₹167,000.00INR / Each
Socorex Acura Adjustable-Volume Pipettor, 1-10mL; Each 2501520 Mfr # W810318
COMPARE
10000 1000
₹154,000.00INR / Each