Silicone-Rubber Heater Blankets Plastic Surfaces with Adhesive-240 VAC
Rugged, moisture / chemical resistant flexible heaters provide uniform heat distribution across large surface areas
  • Maximum exposure temperature of 450°F (232°C)
  • Easy-to-apply industrial strength pressure sensitive adhesive backing
These versatile and industrial-strength heaters can be used on a wide range of surfaces and environments due to their flexibility, moisture / chemical resistance, and ruggedness. They easily install on everything from laboratory vessels to large vats and tanks - ideal for solving viscosity, freeze protection, and process control heating applications. The unique grounded heating element is laminated between two layers of extra strong 20mil fiberglass reinforced silicone rubber. Models for plastic surfaces have a lower 1.25 watts/in2 power density to prevent scorching of the surface
29,200.00 - ₹164,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat SRP06122P Silicone Heating Blanket with Adhesive, for Plastic, 6x12" , 240 V, 90 W 3600155 Mfr # SRP06122P
COMPARE
12 450 240
₹29,200.00INR / Each
BriskHeat SRP06242P Silicone Heating Blanket with Adhesive, for Plastic, 6x24" , 240 V, 180 W 3600159 Mfr # SRP06242P
COMPARE
24 450 240
₹41,100.00INR / Each
BriskHeat SRP12242P Silicone Heating Blanket with Adhesive, for Plastic, 12x24" , 240 V, 360 W 3600167 Mfr # SRP12242P
COMPARE
24 450 240
₹63,000.00INR / Each
BriskHeat SRP24362P Silicone Heating Blanket with Adhesive, for Plastic, 12x36" , 240 V, 1080 W 3600177 Mfr # SRP24362P
COMPARE
36 450 240
₹164,000.00INR / Each