Silicone-Rubber Heater Blankets Metal Surfaces with Adhesive-120 VAC
Rugged, moisture / chemical resistant flexible heaters provide uniform heat distribution across large surface areas
  • Maximum exposure temperature of 450°F (232°C)
  • Easy-to-apply industrial strength pressure sensitive adhesive backing
These versatile and industrial-strength heaters can be used on a wide range of surfaces and environments due to their flexibility, moisture / chemical resistance, and ruggedness. They easily install on everything from laboratory vessels to large vats and tanks - ideal for solving viscosity, freeze protection, and process control heating applications. The unique grounded heating element is laminated between two layers of extra strong 20mil fiberglass reinforced silicone rubber. Heaters for metal surfaces have faster heat-up rates due to the 2.5 watts /in2 power density.
29,200.00 - ₹106,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat SRL06121P Silicone Heating Blanket with Adhesive, for Metal, 6x12" , 120 V, 180 W 3600101 Mfr # SRL06121P
COMPARE
1 450 120
₹29,200.00INR / Each
BriskHeat SRL06241P Silicone Heating Blanket with Adhesive, for Metal, 6x24" , 120 V, 360 W 3600103 Mfr # SRL06241P
COMPARE
2 450 120
₹41,100.00INR / Each
BriskHeat SRL12121P Silicone Heating Blanket with Adhesive, for Metal, 12x12" , 120 V, 360 W 3600108 Mfr # SRL12121P
COMPARE
1 450 120
₹37,200.00INR / Each
BriskHeat SRL12241P Silicone Heating Blanket with Adhesive, for Metal, 12x24" , 120 V, 720 W 3600111 Mfr # SRL12241P
COMPARE
2 450 120
₹63,000.00INR / Each
BriskHeat SRL12361P Silicone Heating Blanket with Adhesive, for Metal, 12x36" , 120 V, 1080 W 3600113 Mfr # SRL12361P
COMPARE
3 450 120
₹106,000.00INR / Each
BriskHeat SRL18181P Silicone Heating Blanket with Adhesive, for Metal, 18x18" , 120 V, 810 W 3600116 Mfr # SRL18181P
COMPARE
1.5 450 120
₹80,300.00INR / Each