Shimpo DT-311D Heavy Duty, AC Powered Digital Xenon Stroboscope
Provides the illusion of “stopped motion”
  • High flash rate of up to 35,000 FPM
  • Manual flash controller knob
  • Highly accurate (±0.01% of reading)
  • Microprocessor-controlled
  • Synchronous output
  • Flash timer control
  • Continuous duty cycle
The DT-311D stroboscope provides the illusion of “stopped motion.” By adjusting the strobe’s flash rate, equipment appears to be standing still or in slow motion. The stroboscope also measures rotational (RPM) or reciprocating (strokes per minute) speeds with the same precision as an electronic digital tachometer.
Ideally suited for printing inspection, motion analysis, preventive maintenance, and speed measurement of repetitive motion. The strobes provide timely data on vibrating parts, leaks, spray patterns and a variety of industrial and scientific operations. It is extremely easy to maneuver and operate via its lightweight and compact design and simplified user interface. Its rugged metal construction provides unequaled reliability in the harshest environments.
226,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Display Type Max Flash Range (fpm) Description
Availability Pricing
Shimpo DT-311D-KIT Heavy Duty, Universally Powered (85-240 VAC) Xenon Stroboscope Kit with Spare Flashtube and Carrying Case 2530531 Mfr # DT-311D-KIT
COMPARE
5 Digit LED 35000 Heavy Duty, Universally Powered (85-240 VAC) Xenon Stroboscope Kit with Spare Flashtube and Carrying Case
₹226,000.00INR / Each