Shimpo DT-3011N Heavy Duty AC Powered Stroboscopes
Provides the illusion of “stopped motion” by adjusting the flash rate
  • Aluminum construction provides exceptional durability
  • High polish and focused reflector eliminates “blind” viewing areas
  • External trigger allows the unit to be automatically synchronized with equipment
  • Light emission auto-off timer (1 -120 min) conserves flash tube life
  • Phase shift, set in degrees or ms, allows visual analysis of rotating/reciprocating objects
  • Save up to 5 user configurations to enable switching quickly between machinery
  • Synchronous output enables master strobe to drive the flash rate of connected strobes
  • Easy tri-pod or permanent mounting with integral mounting thread
By adjusting the flash rate, systems in motion appear to be standing still or in slow motion. These strobes can measure rotational (RPM) or reciprocating (strokes per minute) speeds with the same precision as an electronic digital tachometer. Ideally suited for printing inspection, motion analysis, preventive maintenance, and speed measurement of repetitive motion, this strobe provides timely data on vibrating parts, leaks, spray patterns and a variety of industrial and scientific operations. It is extremely easy to maneuver and operate via its lightweight, compact design combined with a simplified user interface. Its rugged metal construction provides unequaled reliability in the harshest environments.
313,000.00 - ₹374,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Flash Duration Max Flash Range (fpm) Description
Availability Pricing
Shimpo Heavy Duty, AC Powered Stroboscope 2530521 Mfr # DT-3011N
COMPARE
20 μsec 35000 Heavy Duty, AC Powered Stroboscope
₹313,000.00INR / Each
Shimpo Heavy Duty, AC Powered Stroboscope Kit with Spare Flashtube and Carrying Case 2530522 Mfr # DT-3011N-KIT
COMPARE
20 μsec 35000 Heavy Duty, AC Powered Stroboscope Kit with Spare Flashtube and Carrying Case
₹374,000.00INR / Each