Serum Vial Racks
Withstands elevated water temperatures for incubation
  • End plate handles
  • Steam autoclavable at 121 ºC (250 ºF)
These polypropylene racks fit most water baths for elevated temperature incubations. They feature long-lasting construction and are corrosion resistant. Height from base to shelf is 1-5/8" (41 mm). Use these racks with 10-13 mm serum vials, RIA vials, and T3-T4 reaction vials (13-16 mm).
8,100.00 - ₹8,600.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Tube Size (mm) Min Tube Size (mm) Tubes Held
Availability Pricing
Scienceware Serum Vial Rack for 10 to 13-mm Vials WW-06732-00 Mfr # 18513-0072
COMPARE
13 10 72
₹8,600.00INR / Each
Scienceware Serum Vial Rack for 13 to 16-mm Vials WW-06732-10 Mfr # 18513-1640
COMPARE
16 13 40
₹8,100.00INR / Each