Secador® Reusable Silica Gel Desiccant Cartridges
Eliminate the mess of loose desiccant in a reusable design
  • Color indicating desiccant changes color as it absorbs moisture
  • Silica gel absorbs moisture through perforations in the container
  • Reusable desiccant can be easily regenerated
These Bel-Art reusable silica gel desiccant cartridges eliminate the mess of loose desiccant. The silica gel inside the canister absorbs moisture through perforations. Easily view desiccant to determine when it needs regeneration.
Choose from two options—desiccant canister with Desi-Can™ silica gel or desiccant cartridges with silica gel.
The canister maintains a safe level of humidity in 0.09 cubic meter (3 cubic ft.) of space. It is ideal for use in closets, containers, toolboxes, camera cases and other storage areas to prevent rust and moisture. It can be regenerated in a 150º C (300º F) oven in about 3 hours for unlimited life.
The cartridges are highly porous mesh membranes that cover the desiccant and dish, providing excellent absorption. These fit “Space Saver” desiccators, in addition to other plastic desiccators of similar size. Desiccant can be easily regenerated by heating cartridges in a 50°C (122°F) oven overnight or at 100°C (212°F) for 6 hours. For more rapid drying, remove the membrane lid, transfer the spent gel crystals into a glass or porcelain dish, and place in a 150°C (300°F) oven for one hour.
9,600.00 - ₹34,700.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Orientation Description
Availability Pricing
Secador Reusable Desiccant Canister with Desi-Can™ Silica Gel, 4" L x 2" W 0719391 Mfr # 42046-0000
COMPARE
Silica Gel Filled Aluminum Canister Reusable Desiccant Canister with Desi-Can™ Silica Gel, 4" L x 2" W
₹9,600.00INR / Each
Secador Reusable Desiccant Cartridges with Silica Gel, 72 mm Diameter; 6/PK 0719392 Mfr # 420490100
COMPARE
Silica Gel Filled Polypropylene Cartridge Reusable Desiccant Cartridges with Silica Gel, 72 mm Diameter; 6/Pk
₹34,700.00INR / Pkg of 6