Secador® Desiccator Cabinets
Reliable dry storage for humidity sensitive products
  • Large clear polycarbonate panels
  • LCD displays internal temperature and humidity level
  • Three adjustable shelves
These lightweight desiccator cabinets offer dry storage for humidity sensitive products such as laboratory reagents and samples, optical equipment, electronics, and 3D printing filament. The design features high density foam gaskets which keep ambient humidity out and dry air inside. Clear polycarbonate panels allow you to view contents within the cabinets, while the LCD panel located on the outside for the units clearly show the internal temperature and humidity level. Three removable shelves can easily be adjusted within the 1.6 cu ft cabinet interior to meet various height requirements of your products. Additionally shelves can be purchased separately (05006-28).
Two models are available. The electronic model allows you to operate the unit from a 100 to 240 VAC power source without the need of a converter and keeps internal humidity at 10-40%. The non-electronic model can be used with any other Bel-Art desiccant product.
561,000.00 - ₹963,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Chamber Volume (cu ft) Exterior Height (cm) Material
Availability Pricing
Secador 5.0 Desiccator Cabinet 0500627 Mfr # 420570001
COMPARE
1.6 52 Polycarbonate panels & shelves, ABS body
₹561,000.00INR / Each
Secador 5.0 Auto-Desiccator Cabinet 0500629 Mfr # 420571003
COMPARE
1.6 63.5 Polycarbonate panels & shelves, ABS body
₹963,000.00INR / Each