SE International Radiation Alert™ Ranger Radiation Meters
Large sensor for added sensitivity and increased range
  • Sensitive to low levels of alpha, beta, gamma, and x-rays to keep you safe
  • Backlit display allows you to see readings, even in low-light conditions
  • Red light flashes, and alert beeps with each count detected
  • Adjustable alert level and timer
  • Download data, set alarms, and calibrate with free software
Two Ranger models feature extra-large internal or external probe sensors to offer maximum performance and sensitivity in a compact body. Rugged enough to survey the field, with all the features of a lab unit.||The digital display features large digits and selectable backlight—read under any conditions. A red LED flashes and a 70 dB alert beeps with each count detected (may be muted). Icons also make mode and conditions easy to understand.||Save individual readings or timed captures to the internal memory and export to computer through USB with the free software. The software also allows you to set alarm levels and calibrate the meter. Both a cable and software are included.||Built-in efficiencies for Sulfur (S35), Strontium (Sr/y90), Cesium (Cs137), Phosphorus (P32), Carbon (C14), Iodine (I131), Cobalt (Co60), and alpha radiation.
245,000.00 - ₹314,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Sensor Description
Availability Pricing
Radiation Alert Ranger    Digital Radiation Meter with Internal Sensor 8191021 Mfr # RANGER
COMPARE
Internal Digital Radiation Meter with Internal Sensor
₹245,000.00INR / Each
Radiation Alert Ranger EXP Digital Radiation Meter with External Probe 8191023 Mfr # RANGER EXP
COMPARE
External Digital Radiation Meter with External Probe
₹314,000.00INR / Each