SciMatCo Polypropylene Acid Storage Cabinets
Liquid tight to prevent leakage
These polypropylene acid storage cabinets contain no metal parts— they won’t corrode or rust. Constructed of 3/8" (0.96 cm)-thick white polypropylene, cabinets feature plastic hinges and welded interior seams to prevent leakage. All cabinets include hazard warning label.
Compliance: OSHA, UFC, IFC
116,000.00 - ₹643,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Number of Shelves
Availability Pricing
SciMatCo SC5000 Polypropylene Acid Storage Cabinet, 6 X 2.5 L bottle capacity WW-81780-15 Mfr # SC5000
COMPARE
4 0
₹116,000.00INR / Each
SciMatCo SC5060 Polypropylene Acid Storage Cabinet, 100 X 2.5 L bottle capacity WW-81780-42 Mfr # SC5060
COMPARE
66 3
₹643,000.00INR / Each