SciMatCo Flammable Storage Cabinets
Wooden cabinets provide excellent thermal insulation in case of fire
Use this lockable, wooden cabinet to conveniently store flammable liquids. Constructed of 1" (2.5 cm) thick exterior grade plywood; finished with yellow epoxy paint inside and out. Specially designed floor serves as a liquid-tight trough to retain spills. Locking door features continuous piano-type hinges, and a 8" x 8" (20.3 x 20.3 cm) "Flammable" hazard label. Contains one variable position shelf. Meets all applicable OSHA and NFPA standards for flammable safety cabinets. UL Listed.
248,000.00 - ₹369,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Self-Closing
Availability Pricing
SciMatCo SC8023 Flammable Storage Cabinet, 6 gallon capacity, self-closing door 8178025 Mfr # SC8023
COMPARE
6 Yes
₹248,000.00INR / Each
SciMatCo SC8079 Flammable Storage Cabinet, 24 gallon capacity, self-closing door 8178027 Mfr # SC8079
COMPARE
24 Yes
₹369,000.00INR / Each