Scienceware Self Adhesive Waste Bags, HDPE
Hang anywhere for easy waste collection
These versatile bags can be hung from walls, cabinets or anywhere to free up valuable bench space. Simply peel the back off the self-adhesive strip and hang bag near your work area. The longer back panel makes the bag easy to open and use. Made from high-molecular weight HDPE, these white bags are steam autoclavable at 250°F (121°C). Passes both ASTM 1709 Dart Impact and ASTM 1922 Tear Resistance tests. Maximum content weight should not exceed 3lb (1.4 kg).
9,900.00 - ₹16,500.00INR / Pkg of 50
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Width (in)
Availability Pricing
Scienceware F131741008 Self Adhesive Bags, 8" x 10", 50 Pack 0649761 Mfr # F131741008
COMPARE
10 8
₹9,900.00INR / Pkg of 50
Scienceware F131741012 Self Adhesive Bags, 12" x 16", 50 Pack 0649762 Mfr # F131741012
COMPARE
16 12
₹16,500.00INR / Pkg of 50