Scienceware Roxy M.™ Repeating Pipette Tips
Universal fit!
  • Fit most manual repeating pipettors including Roxy M™, Eppendorf® Multipette® 4780, Brand HandyStep® S, Fisherbrand® Repeating Pipetter, VWR® Repeating Pipettor, Gilson Distriman®, and Scilogex® StepMate Repeater Pipettor
Roxy M. nonsterile and individually wrapped sterile tips are nearly universal and fit most manual repeating pipettors. Sized to fit all applications, they indicate the capacity and the repetitive volumes each tip will dispense. A polyethylene inner plunger prevents the liquid from contacting any part of the pipette, eliminating the risk for contamination.
12,100.00 - ₹43,500.00INR / Pkg of 25
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Package Of
Availability Pricing
Scienceware Roxy M. Non-Sterile Repeating Pipettor Tip, 0.05 mL, 100/pk WW-07897-60 Mfr # F373000000
COMPARE
50 Bulk Packed
₹36,000.00INR / Pkg of 100
Scienceware Roxy M. Non-Sterile Repeating Pipettor Tip, 0.5 mL, 100/pk WW-07897-62 Mfr # F373000001
COMPARE
500 Bulk Packed
₹33,100.00INR / Pkg of 100
Scienceware Roxy M. Non-Sterile Repeating Pipettor Tip, 1.25 mL, 100/pk WW-07897-64 Mfr # F373000002
COMPARE
1250 Bulk Packed
₹33,700.00INR / Pkg of 100
Scienceware Roxy M. Non-Sterile Repeating Pipettor Tip, 2.5 mL, 100/pk WW-07897-66 Mfr # F373000003
COMPARE
2500 Bulk Packed
₹33,700.00INR / Pkg of 100
Scienceware Roxy M. Non-Sterile Repeating Pipettor Tip, 5.0 mL, 100/pk WW-07897-68 Mfr # F373000004
COMPARE
5000 Bulk Packed
₹32,800.00INR / Pkg of 100
Scienceware Roxy M. Non-Sterile Repeating Pipettor Tip, 12.5 mL, 100/pk WW-07897-70 Mfr # F373000005
COMPARE
12500 Bulk Packed
₹31,700.00INR / Pkg of 100
Scienceware Roxy M. Non-Sterile Repeating Pipettor Tip, 25 mL, 25/pk WW-07897-72 Mfr # F373000008
COMPARE
25000 Bulk Packed
₹12,100.00INR / Pkg of 25
Scienceware Roxy M. Non-Sterile Repeating Pipettor Tip, 50 mL, 25/pk WW-07897-74 Mfr # F373000009
COMPARE
50000 Bulk Packed
₹13,600.00INR / Pkg of 25
Scienceware Roxy M. Sterile Repeating Pipettor Tip, 0.5 mL, 100/pk WW-07897-78 Mfr # F373000010
COMPARE
500 Individually wrapped (sterile)
₹43,500.00INR / Pkg of 100
Scienceware Roxy M. Sterile Repeating Pipettor Tip, 1.25 mL, 100/pk WW-07897-80 Mfr # F373000011
COMPARE
1250 Individually wrapped (sterile)
₹43,500.00INR / Pkg of 100
Scienceware Roxy M. Sterile Repeating Pipettor Tip, 2.5 mL, 100/pk WW-07897-82 Mfr # F373000012
COMPARE
2500 Individually wrapped (sterile)
₹43,500.00INR / Pkg of 100
Scienceware Roxy M. Sterile Repeating Pipettor Tip, 5.0 mL, 100/pk WW-07897-84 Mfr # F373000013
COMPARE
5000 Individually wrapped (sterile)
₹43,500.00INR / Pkg of 100
Scienceware Roxy M. Sterile Repeating Pipettor Tip, 12.5 mL, 100/pk WW-07897-86 Mfr # F373000014
COMPARE
12500 Individually wrapped (sterile)
₹43,500.00INR / Pkg of 100
Scienceware Roxy M. Sterile Repeating Pipettor Tip, 25 mL, 25/pk WW-07897-88 Mfr # F373000015
COMPARE
25000 Individually wrapped (sterile)
₹15,200.00INR / Pkg of 25
Scienceware Roxy M. Sterile Repeating Pipettor Tip, 50 mL, 25/pk WW-07897-90 Mfr # F373000016
COMPARE
50000 Individually wrapped (sterile)
₹14,900.00INR / Pkg of 25
Scienceware Roxy M. Assorted Repeating Pipettor Tip Pack; 45/pk WW-07897-94 Mfr # F373000100
COMPARE
5000 Assorted Sizes
₹32,800.00INR / Pkg of 45
Showing 1- 16 of 16