Scienceware Portable Glove Boxes
Safe viewing with operator and external environment protection
  • Lightweight, self-contained, portable
  • LDPE-molded body, rounded corners, easy cleaning
  • Large acrylic window, 22" long (20" installed)
  • 6" circular glove ports for safety
  • 8" diameter side port permit easy insertion of material/equipment
  • Neoprene gloves (included) protect against gas, liquid or solid penetration
  • Available with or without gas ports
505,000.00 - ₹538,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Chamber Depth (in) Chamber Height (in) Chamber Width (in)
Availability Pricing
Scienceware H50028-2000 LDPE Glove Box; Acrylic Window; 20" long, 6" Wide Neoprene Gloves, No Gas Port 3477000 Mfr # H50028-2000
COMPARE
27 22 13
₹505,000.00INR / Each
Scienceware H50028-2001 Portable Glove Box; 86.6 L, Neoprene Gloves, 2 Gas Ports 3477002 Mfr # H50028-2001
COMPARE
27 22 13
₹538,000.00INR / Each