Scienceware® Polypropylene Test Tube Racks
Hold samples in baths effectively
  • Open rack drains fluids quickly
  • Models 06737-10, -15, and -20 stack with ease
  • Autoclavable
These racks will not seal or stick in a water bath—open rack design helps drain off fluid. Autoclavable.
7,000.00 - ₹10,900.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Tube Size (mm) Tubes Held
Availability Pricing
Scienceware 18860-1013 Test Tube Rack, PP, for 10 to 13 mm OD Tubes WW-06737-10 Mfr # 18860-1013
COMPARE
13 72
₹7,000.00INR / Each
Scienceware 18860-1316 Test Tube Rack, PP, for 13 to 16 mm OD Tubes WW-06737-15 Mfr # 18860-1316
COMPARE
16 40
₹7,100.00INR / Each
Scienceware 18860-1620 Test Tube Rack, PP, for 16 to 20 mm OD Tubes WW-06737-20 Mfr # 18860-1620
COMPARE
20 40
₹9,200.00INR / Each
Scienceware 18860-2125 Test Tube Rack, PP, for 21 to 25 mm OD Tubes WW-06737-30 Mfr # 18860-2125
COMPARE
25 40
₹10,900.00INR / Each
Scienceware 18860-2630 Test Tube Rack, PP, for 26 to 30 mm OD Tubes WW-06737-40 Mfr # 18860-2630
COMPARE
30 24
₹10,400.00INR / Each