Scienceware® Graduated Tapered Polypropylene Beakers
Tapered beaker design with spout makes for easy pouring
  • Easy stacking and storage
  • Autoclavable
These polypropylene (PP) beakers are tapered for easy stacking and storage. Graduations are engraved into the side of the beaker. Beakers can withstand temperatures to 250°F (121°C).
4,800.00 - ₹12,700.00INR / Pkg of 6
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Scienceware 26211-0000 Tapered Polypropylene Beaker, 100 mL WW-06003-21 Mfr # 26211-0000
COMPARE
100 10 Polypropylene(PP)
₹6,700.00INR / Pkg of 12
Scienceware 26212-0000 Tapered Polypropylene Beaker, 150 mL WW-06003-31 Mfr # 26212-0000
COMPARE
150 25 Polypropylene(PP)
₹12,700.00INR / Pkg of 12
Scienceware 26213-0000 Tapered Polypropylene Beaker, 250 mL WW-06003-41 Mfr # 26213-0000
COMPARE
250 50 Polypropylene(PP)
₹6,000.00INR / Pkg of 6
Scienceware 26214-0000 Tapered Polypropylene Beaker, 400 mL WW-06003-51 Mfr # 26214-0000
COMPARE
400 100 Polypropylene(PP)
₹6,500.00INR / Pkg of 6
Scienceware 26215-0000 Tapered Polypropylene Beaker, 600 mL WW-06003-61 Mfr # 26215-0000
COMPARE
600 100 Polypropylene(PP)
₹6,300.00INR / Pkg of 4
Scienceware 26216-0000 Tapered Polypropylene Beaker, 1000 mL WW-06003-71 Mfr # 26216-0000
COMPARE
1000 200 Polypropylene(PP)
₹4,800.00INR / Pkg of 3