Scienceware Dippers
Easily pour from either side
  • Long handles for easy sampling
Lightweight, yet sturdy, dippers feature a polypropylene (PP) cup that screws onto the handle — order extra cups for your multiple sampling applications. Two pouring spouts on the cup allow you to pour from either side. Long handles make these dippers ideal for sampling from large tanks, vats, streams, and lakes. High-density polyethylene (HDPE) handles are ribbed for a sure grip; end has hook for hanging.
20,300.00 - ₹26,500.00INR / Pkg of 1
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Handle Length (InchShort) Sterile
Availability Pricing
Scienceware F36784-0016 Polypropylene Dipper, 16 oz, 4" dia bowl, 36"L HDPE handle 0628200 Mfr # F36784-0016
COMPARE
500 36 Yes
₹20,300.00INR / Pkg of 1
Scienceware F36785-0016 Polypropylene Dipper, 16 oz, 4" dia bowl, 72"L HDPE handle 0628205 Mfr # F36785-0016
COMPARE
500 72 Yes
₹26,500.00INR / Pkg of 1