Savillex Lab Bottles with Closure, PFA
Ultra-pure PFA bottles with superior temperature resistance for critical applications
  • PFA material provides excellent chemical resistance
  • Withstands temperature as low as –200°C (–328°F)
  • Stretch blow molded for smooth surface finish
  • Leakproof PFA screw
  • Manufactured and sealed in bags inside a cleanroom
Savillex PFA lab bottles deliver unmatched chemical purity, durability, and a wide temperature range for demanding applications. Manufactured using a unique stretch blow molding process, these high-performance bottles offer lower trace metal content, smoother surface finish, easier pouring, and superior sealing. The linerless PFA screw closures provide a secure, leak-free seal. Working temperature range is from -200 to 260°C (-328 to 500°F)
10,500.00 - ₹58,400.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size
Availability Pricing
Savillex Lab Bottle with 33 mm Closure with Lugs, PFA, 50 mL  0605200 Mfr # 150-01-0050
COMPARE
50 33mm
₹10,500.00INR / Each
Savillex Lab Bottle with GL45 Closure, PFA, 100 mL  0605201 Mfr # 150-01-0100
COMPARE
100 GL45
₹11,900.00INR / Each
Savillex Lab Bottle with GL45 Closure, PFA, 250 mL 0605202 Mfr # 150-01-0250
COMPARE
250 GL45
₹17,000.00INR / Each
Savillex Lab Bottle with GL45 Closure, PFA, 500 mL 0605203 Mfr # 150-01-0500
COMPARE
500 GL45
₹23,800.00INR / Each
Savillex Lab Bottle with GL45 Closure, PFA, 1000 mL 0605204 Mfr # 150-01-1000
COMPARE
1000 GL45
₹37,700.00INR / Each
Savillex Lab Bottle with GL45 Closure, PFA, 2000 mL  0605205 Mfr # 150-01-2000
COMPARE
2000 GL45
₹58,400.00INR / Each