Sartorius Entris® II NTEP-Approved Analytical Balances
High accuracy and fast stabilization time ensure excellent weighing result every time
  • NTEP-approved legal-for-trade balances offer quality precision weighing
  • LED touch technology and 12 built-in applications
  • Constructed of highly chemical-resistant materials
Sartorius Entris® II analytical balances offer a better accuracy than their top-loading counterparts. Specially coated glass draft shield reduces weighing errors caused by electrostatically charged samples. Constructed of durable polybutylene terephthalate (PBT), stainless steel, and glass, these balances provide excellent chemical resistance and are easy to clean. Quick wipe down design and easy-to-remove parts help prevent cross contamination.
Built-in applications such as dosing, counting, percentage weighing, mixing, totalizing, animal weighing, free factor, density determination, statistics, peakhold, checkweighing, mass unit conversion, and underfloor weighing help make your weighing process more efficient. Easily connect to your PC using the RS-232 or USB-C interfaces. They are “PC-direct”, meaning no extra software is required and you can download your data directly to Microsoft® Word or Excel.
593,000.00 - ₹669,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (mg)
Availability Pricing
Sartorius BCE124-1NUS Entris® II NTEP-Approved Analytical Balance, 120 g x 0.1 mg 5500027 Mfr # BCE124-1NUS
COMPARE
120 0.1
₹593,000.00INR / Each
Sartorius BCE224-1NUS Entris® II NTEP-Approved Analytical Balance, 220 g x 0.1 mg 5500028 Mfr # BCE224-1NUS
COMPARE
220 0.1
₹669,000.00INR / Each