Sartorius® Cubis® II (MCA) High-Capacity Micro Analytical Balances without Software
Offers outstanding accuracy and repeatability with the flexibility to add software and upgrade hardware
  • Scroll and swipe through applications on the large 7" color touch display
  • Error-free operation with outstanding accuracy and repeatability
  • Ultra-high resolution with up to 61 million weighing steps
  • End-to-end data integrity
  • 21 CFR Part 11 and EU Annex 11 compliance
  • Integrated audit-trail
  • Motorized auto-leveling
  • State-of-the-art user management
  • Innovative TFT touch screen operation
  • Tool-free assembly/disassembly for easy cleaning
  • High chemical resistance to most cleaning agents
These high-capacity balances feature an easy-to-read, large, high-contrast TFT touch screen with clearly structured functionality and essential weighing applications. The balances are simple to operate and allow you to weigh very small samples directly into large flasks with the highest level of accuracy and repeatability.
The balances are less prone to environmental effects, resulting in faster weighing workflows and improved performance in a daily laboratory environment. A built-in intelligent compensating system manages temperature, humidity, and air pressure changes which further ensures stable balance behavior. A spacious weighing area supports ergonomic weighing into various flasks.
The MCA model balances are easily adaptable to your specific laboratory needs. Various software packages are available to enhance your measuring application; select to add Pharma QP1 (compliance with pharmaceutical relevant guidelines), Advanced Applications QP2 (complex weighing applications), Utilities QP3 (helpful tools), Connectivity QP4 (connectivity for data exchange), or Hardware QP10 (licensable hardware functions). Models are available with a built-in ionizer for samples prone to static charges and automated draft shield for hands-free convenience. If you choose to order models without those features at this time, you can simply add an ionizer or an automated draft shield later at your convenience.
9,644,000.00 - ₹16,281,000.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Repeatability (mg) Description
Availability Pricing
Sartorius MCA36S-3S00-D HWL Cubis® II High-Capacity Micro Balance, 32g x 0.001mg; Activated Ionizer, Motorized Draft Shield 3974315 Mfr # MCA36S-3S00-D HWL
COMPARE
32 0.0015 High-Capacity Micro Balance, 32g x 0.001mg; Activated Ionizer, Motorized Draft Shield
₹13,402,000.00INR / Each
Sartorius MCA36S-3S00-D Cubis® II High-Capacity Micro Balance, 32g x 0.001mg; Manual Draft Shield 3974316 Mfr # MCA36S-3S00-D
COMPARE
32 0.0015 High-Capacity Micro Balance, 32g x 0.001mg; Manual Draft Shield
₹13,085,000.00INR / Each
Sartorius MCA36P-3S00-D HWL Cubis® II High-Capacity Micro Balance, 10g/32g x 0.001mg/0.01mg; Activated Ionizer, Motorized Draft Shield 3974317 Mfr # MCA36P-3S00-D HWL
COMPARE
32 0.002 High-Capacity Micro Balance, 10g/32g x 0.001mg/0.01mg; Activated Ionizer, Motorized Draft Shield
₹11,313,000.00INR / Each
Sartorius MCA36P-3S00-D Cubis® II High-Capacity Micro Balance, 10g/32g x 0.001mg/0.01mg; Manual Draft Shield 3974318 Mfr # MCA36P-3S00-D
COMPARE
32 0.002 High-Capacity Micro Balance, 10g/32g x 0.001mg/0.01mg; Manual Draft Shield
₹10,996,000.00INR / Each
Sartorius MCA66S-3S00-D HWL Cubis® II High-Capacity Micro Balance, 61g x 0.001mg; Activated Ionizer, Motorized Draft Shield 3974319 Mfr # MCA66S-3S00-D HWL
COMPARE
61 0.0015 High-Capacity Micro Balance, 61g x 0.001mg; Activated Ionizer, Motorized Draft Shield
₹16,281,000.00INR / Each
Sartorius MCA66S-3S00-D Cubis® II High-Capacity Micro Balance, 61g x 0.001mg; Manual Draft Shield 3974320 Mfr # MCA66S-3S00-D
COMPARE
61 0.0015 High-Capacity Micro Balance, 61g x 0.001mg; Manual Draft Shield
₹15,964,000.00INR / Each
Sartorius MCA66P-3S00-D HWL Cubis® II High-Capacity Micro Balance, 12g/61g x 0.001mg/0.01mg; Activated Ionizer, Motorized Draft Shield 3974321 Mfr # MCA66P-3S00-D HWL
COMPARE
61 0.002 High-Capacity Micro Balance, 12g/61g x 0.001mg/0.01mg; Activated Ionizer, Motorized Draft Shield
₹15,044,000.00INR / Each
Sartorius MCA66P-3S00-D Cubis® II High-Capacity Micro Balance, 12g/61g x 0.001mg/0.01mg; Manual Draft Shield 3974322 Mfr # MCA66P-3S00-D
COMPARE
61 0.002 High-Capacity Micro Balance, 12g/61g x 0.001mg/0.01mg; Manual Draft Shield
₹14,727,000.00INR / Each
Sartorius MCA116S-3S00-D HWL Cubis® II High-Capacity Micro Balance, 111g x 0.002mg; Activated Ionizer, Motorized Draft Shield 3974323 Mfr # MCA116S-3S00-D HWL
COMPARE
111 0.004 High-Capacity Micro Balance, 111g x 0.002mg; Activated Ionizer, Motorized Draft Shield
₹9,962,000.00INR / Each
Sartorius MCA116S-3S00-D Cubis® II High-Capacity Micro Balance, 111g x 0.002mg; Manual Draft Shield 3974324 Mfr # MCA116S-3S00-D
COMPARE
111 0.004 High-Capacity Micro Balance, 111g x 0.002mg; Manual Draft Shield
₹9,644,000.00INR / Each
Showing 1- 10 of 10